(3S)-1-Chloro-3-tosylamido-7-amino-2-heptanone hydrochloride 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: TLCK, Nalpha-p-Tosyl-L-lysine chloromethyl ketone hydrochloride, Tos-Lys-chloromethylketone·HCl

22795-0010 22795-0500 22795-2500
TS22795-0010EA 834 USD
TS22795-0010 TS22795-0500 TS22795-2500
(3S)-1-Chloro-3-tosylamido-7-amino-2-heptanone hydrochloride 98%
(3S)-1-Chloro-3-tosylamido-7-amino-2-heptanone hydrochloride
Formula: C₁₄H₂₂Cl₂N₂O₃S
MW: 369.301 g/mol
Melting Pt: 160…161 °C (dec.)
Storage Temperature: Refrigerator
MDL Number: MFCD00065395
CAS Number: 4272-74-6

Order Now

Specification Test Results

Appearance White to light beige Powder
Infrared spectrum Conforms
Titration Argentometric ≥97.5 %
Appearance of solution (5% soln. in methanol) clear, slightly colored
Specific optical rotation -9° to -10° (24°C, 589 nm) (c=5, H2O)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR