N-(p-Toluenesulfonyl)-L-phenylalanylchloromethylketone 99+%
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
L-1-4'-Tosylamino-2-phenylethyl chloromethyl ketone, N-Tosyl-L-phenylalanyl Chloromethyl Ketone, (S)-N-(4-Chloro-3-oxo-1-phenyl-2-butyl)-p-toluenesulphonamide, TPCK
22780-0010
22780-0050
TS22780-0010EA
171
USD
TS22780-0010
TS22780-0050
N-(p-Toluenesulfonyl)-L-phenylalanylchloromethylketone 99+%
N-(p-Toluenesulfonyl)-L-phenylalanylchloromethylketone
Formula:
C₁₇H₁₈ClNO₃S Melting Pt: 105…109 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00000935 CAS Number: 402-71-1 |
Specification Test Results
Appearance | White Crystalline powder |
Infrared spectrum | Conforms |
Melting point | 105 to 109 °C |
TLC | ≥98.0 % |
Specific optical rotation | -86° to -88° (20 °C, 589 nm) (c=1, abs. ethanol) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...