Methyl-4-formylbenzoate 99%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Methyl terephthalaldehydate, Methyl 4-formylbenzoate

22375-0100 22375-0500
TS22375-0100EA 115 USD
TS22375-0100 TS22375-0500
Methyl-4-formylbenzoate 99%
Methyl-4-formylbenzoate
Formula: C₉H₈O₃
MW: 164.16 g/mol
Melting Pt: 57…64 °C
Flash Pt: 144 °C
Storage Temperature: Ambient
MDL Number: MFCD00006950
CAS Number: 1571-08-0
EINECS: 216-385-5

Order Now

Specification Test Results

Appearance White to cream to light green Crystalline powder and/or chunks
Infrared spectrum Conforms
Melting point 59 to 63 °C
GC ≥98.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR