2-Butyne-1-ol 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 2-Butyn-1-ol, But-2-yn-1-ol, 1-Hydroxy-2-butyne

22340-0050 22340-0250
TS22340-0050EA 65 USD
TS22340-0050 TS22340-0250
2-Butyne-1-ol 98%
2-Butyne-1-ol
Formula: C₄H₆O
MW: 70.09 g/mol
Boiling Pt: 142
Melting Pt: -2
Density: 0.93 g/cm³
Flash Pt: 51 °C
Storage Temperature: Ambient
MDL Number: MFCD00002914
CAS Number: 764-01-2
EINECS: 212-113-4
UN: 1987
ADR: 3 III

Order Now

Specification Test Results

Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.4520 to 1.4540 (20 °C, 589 nm)
GC ≥97.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR