Bromoacetaldehyde dimethyl acetal 97% stabilised

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: 2-Bromo-1,1-dimethoxyethane

22187-0250 22187-1000
TS22187-0250EA 50 USD
TS22187-0250 TS22187-1000
Bromoacetaldehyde dimethyl acetal 97% stabilised
Bromoacetaldehyde dimethyl acetal
Formula: BrH₂CCH(OCH₃)₂
MW: 169.02 g/mol
Boiling Pt: 148
Density: 1.43 g/cm³
Flash Pt: 51 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00000213
CAS Number: 7252-83-7
EINECS: 230-669-6
UN: 2929
ADR: 6.1 II

Order Now

Specification Test Results

Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.4450 to 1.4470 (20 °C, 589 nm)
GC ≥96.0 %
Stabilizer 0.2 % potassium carbonate


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR