2,3-Pentanedione 97%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Pentane-2,3-dione

22147-0250 22147-1000 22147-5000
TS22147-0250EA 38 USD
TS22147-0250 TS22147-1000 TS22147-5000
2,3-Pentanedione 97%
2,3-Pentanedione
Formula: C₅H₈O₂
MW: 100.12 g/mol
Boiling Pt: 110…112 °C (760 mmHg)
Melting Pt: –52 °C
Density: 0.958 g/cm³
Flash Pt: 18 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00009313
CAS Number: 600-14-6
EINECS: 209-984-8
UN: 1224
ADR: 3,II

Order Now

Specification Test Results

Appearance Clear yellow Liquid
Infrared spectrum Conforms
Refractive index 1.4020 to 1.4060 (20 °C, 589 nm)
GC ≥96.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR