(R)-(-)-2,2-Dimethyl-1,3-dioxolan-4-ylmethanol 98%

Supplier: Thermo Scientific Chemicals

Synonyms: (R)-(-)-1,2-O-Isopropylidenglycerol, (R)-(-)-2,2-Dimethyl-1,3-dioxolane-4-methanol, (R)-(-)-2,3-O-Isopropylideneglycerol, (R)-(-)-1,2-O-Isopropylideneglycerol, (R)-(2,2-Dimethyl-1,3-dioxolan-4-yl)methanol

22133-0010 22133-0050
TS22133-0010EA 43 USD
TS22133-0010 TS22133-0050
(R)-(-)-2,2-Dimethyl-1,3-dioxolan-4-ylmethanol 98%
(R)-(-)-2,2-Dimethyl-1,3-dioxolan-4-ylmethanol
Formula: C₆H₁₂O₃
MW: 132.16 g/mol
Boiling Pt: 72…73 °C (8 mmHg)
Density: 1.06 g/cm³
Flash Pt: 80 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00003213
CAS Number: 14347-78-5
EINECS: 405-320-8
Merck Index: 12,05232

Order Now

Specification Test Results

Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.4330 to 1.4360 (20 °C, 589 nm)
GC ≥97.5 %
Specific optical rotation -13° to -15° (20°C, 589 nm) (neat)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR