1,1,1,5,5,5-Hexafluoroacetylacetone 99%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Hexafluoroacetylacetone, 1,1,1,5,5,5-Hexafluoropentane-2,4-dione, 1,1,1,5,5,5-Hexafluoro-2,4-pentanedione, HFAA

21853-0050 21853-0250 21853-1000
TS21853-0050EA 59 USD
TS21853-0050 TS21853-0250 TS21853-1000
1,1,1,5,5,5-Hexafluoroacetylacetone 99%
1,1,1,5,5,5-Hexafluoroacetylacetone
Formula: C₅H₂F₆O₂
MW: 208.06 g/mol
Boiling Pt: 70…71 °C (760 mmHg)
Density: 1.47 g/cm³
Flash Pt: 32 °C
Storage Temperature: Ambient
MDL Number: MFCD00000426
CAS Number: 1522-22-1
EINECS: 216-191-0
UN: 2920
ADR: 8,II

Order Now

Specification Test Results

Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.3320 to 1.3340 (20 °C, 589 nm)
GC ≥98.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR