(S)-(-)-1-Phenylethanol 99%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: (S)-(-)-α-Methylbenzyl alcohol, (S)-(-)-sec-Phenethyl alcohol, (S)-1-Phenylethanol

21779-0010 21779-0050 21779-0250
TS21779-0010EA 56 USD
TS21779-0010 TS21779-0050 TS21779-0250
(S)-(-)-1-Phenylethanol 99%
(S)-(-)-1-Phenylethanol
Formula: C₈H₁₀O
MW: 122.17 g/mol
Boiling Pt: 98 °C (20 mmHg)
Melting Pt: 9…11 °C
Density: 1.01 g/cm³
Flash Pt: 85 °C
Storage Temperature: Ambient
MDL Number: MFCD00064264
CAS Number: 1445-91-6
UN: 2937
ADR: 6.1,III

Order Now

Specification Test Results

Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.5260 to 1.5280 (20 °C, 589 nm)
GC ≥98.5 %
Specific optical rotation -41° to -44° (20°C, 589 nm) (neat)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR