Disodium fumarate 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Disodium Fumarate, Fumaric acid disodium salt, Fumaric Acid Disodium Salt, Sodium fumarate

21553-0050 21553-1000 21553-5000
TS21553-0050EA 23 USD
TS21553-0050 TS21553-1000 TS21553-5000
Disodium fumarate 98%
Disodium fumarate
Formula: C₄H₂Na₂O₄
MW: 160.04 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00064567
CAS Number: 17013-01-3
EINECS: 241-087-7

Order Now

Specification Test Results

Appearance White Crystalline powder
Infrared spectrum Conforms
Titration with HClO4 ≥97.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR