Arachidyl alcohol 98%

Supplier: Thermo Scientific Chemicals

21546-0050 21546-0250
TS21546-0050EA 36 USD
TS21546-0050 TS21546-0250
Arachidyl alcohol 98%
Arachidyl alcohol
Formula: H₃C(CH₂)₁₈CH₂OH
Melting Pt: 64…66 °C
Flash Pt: 194 °C
Storage Temperature: Ambient
MDL Number: MFCD00002938
CAS Number: 629-96-9
EINECS: 211-119-4

Order Now

Specification Test Results

Appearance White Powder or pellets and/or chunks
Infrared spectrum Conforms
Melting point 62 to 66 °C
GC ≥97.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR