4'-(Trifluoromethyl)acetophenone 99%

Supplier: Thermo Scientific Chemicals
Warning

21445-0010 21445-0050 21445-0250
TS21445-0010EA 35 USD
TS21445-0010 TS21445-0050 TS21445-0250
4'-(Trifluoromethyl)acetophenone 99%
4'-(Trifluoromethyl)acetophenone
Formula: C₉H₇F₃O
Boiling Pt: 79…80 °C (8 mmHg)
Melting Pt: 30…33 °C
Density: 0.92 g/cm³
Flash Pt: 84 °C
Storage Temperature: Ambient
MDL Number: MFCD00000401
CAS Number: 709-63-7
EINECS: 211-913-0
UN: 1325
ADR: 4.1,II

Order Now

Specification Test Results

Appearance White to light yellow Low melting solid
Infrared spectrum Conforms
Melting point 30 to 33 °C
GC ≥98.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR