3-Ethoxymethacrolein 95% tech.

Supplier: Thermo Scientific Chemicals
Warning

21399-0010 21399-0100
TS21399-0010EA 58 USD
TS21399-0010 TS21399-0100
3-Ethoxymethacrolein 95% tech.
3-Ethoxymethacrolein
Formula: C₆H₁₀O₂
Boiling Pt: 78…81 °C (14 mmHg)
Density: 0.96 g/cm³
Flash Pt: 35 °C
Storage Temperature: Refrigerator
CAS Number: 42588-57-8
UN: 3271
ADR: 3,III

Order Now

Specification Test Results

Appearance Colorless to yellow Liquid
Infrared spectrum Conforms
Refractive index 1.4775 to 1.4815 (20 °C, 589 nm)
GC ≥94 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR