2-(2-Bromoethyl)-1,3-dioxolane 96%

Supplier: Thermo Scientific Chemicals
Warning

21354-0100 21354-0250 21354-0500
TS21354-0100EA 61 USD
TS21354-0100 TS21354-0250 TS21354-0500
2-(2-Bromoethyl)-1,3-dioxolane 96%
2-(2-Bromoethyl)-1,3-dioxolane
Formula: C₅H₉BrO₂
MW: 181.03 g/mol
Boiling Pt: 68…70 °C (8 mmHg)
Density: 1.54 g/cm³
Flash Pt: 65 °C
Storage Temperature: Freezer
MDL Number: MFCD00003216
CAS Number: 18742-02-4
EINECS: 242-551-1

Order Now

Specification Test Results

Appearance Clear colorless to yellow to brown Liquid
Infrared spectrum Conforms
Refractive index 1.4780 to 1.4830 (20 °C, 589 nm)
GC ≥95 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR