5-Hexen-1-ol 99%

Supplier: Thermo Scientific Chemicals
Warning

21304-0050 21304-0250 21304-1000
TS21304-0050EA 75 USD
TS21304-0050 TS21304-0250 TS21304-1000
5-Hexen-1-ol 99%
5-Hexen-1-ol
Formula: C₆H₁₂O
Boiling Pt: 152…155 °C (760 mmHg)
Density: 0.8725 g/cm³
Flash Pt: 47 °C
Storage Temperature: Ambient
MDL Number: MFCD00002981
CAS Number: 821-41-0
EINECS: 212-477-4
UN: 1987
ADR: 3,III

Order Now

Specification Test Results

Appearance Clear colorless Liquid
Infrared spectrum Conforms
Refractive index 1.4340 to 1.4360 (20 °C, 589 nm)
GC ≥98.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR