Aluminium isopropoxide 99.99% (trace metals basis)

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Aluminiumtriisopropylate, Aluminiumisopropylate, Aluminiumtriisopropoxide, Aluminium isopropoxide

21223-0100 21223-0500 21223-2500
TS21223-0100EA 55 USD
TS21223-0100 TS21223-0500 TS21223-2500
Aluminium isopropoxide 99.99% (trace metals basis)
Aluminium isopropoxide
Formula: C₉H₂₁AlO₃
MW: 204.25 g/mol
Boiling Pt: 135
Melting Pt: 119
Storage Temperature: Ambient
MDL Number: MFCD00008870
CAS Number: 555-31-7
EINECS: 209-090-8
UN: 3181
ADR: 4.1 II
Merck Index: 13,00346

Order Now

Specification Test Results

Appearance White Powder and/or chunks
Infrared spectrum Conforms
Total trace metal impurities ≤150 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR