ω,ω-Diethoxyacetophenone 96%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 2,2-Diethoxyacetophenone, 2,2-Diethoxy-1-phenylethan-1-one

21062-0050 21062-1000
TS21062-0050EA 24 USD
TS21062-0050 TS21062-1000
ω,ω-Diethoxyacetophenone 96%
ω,ω-Diethoxyacetophenone
Formula: C₁₂H₁₆O₃
MW: 208.26 g/mol
Boiling Pt: 131…134 °C (10 mmHg)
Density: 1.03 g/cm³
Flash Pt: 128 °C
MDL Number: MFCD00009659
CAS Number: 6175-45-7
EINECS: 228-220-4

Order Now

Specification Test Results

Appearance Clear colorless to dark yellow Liquid
Infrared spectrum Conforms
Refractive index 1.4980 to 1.5010 (20 °C, 589 nm)
GC ≥95 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR