2-(Bromomethyl)-1,3-dioxolane 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
2-Bromomethyl-1,3-dioxolane
20984-0250
20984-1000
TS20984-0250EA
78
USD
TS20984-0250
TS20984-1000
2-(Bromomethyl)-1,3-dioxolane 97%
2-(Bromomethyl)-1,3-dioxolane
Formula:
C₄H₇BrO₂ MW: 167 g/mol Boiling Pt: 80…82 °C (27 mmHg) Flash Pt: 62 °C Storage Temperature: Ambient |
MDL Number:
MFCD00003214 CAS Number: 4360-63-8 EINECS: 224-443-6 |
Specification Test Results
Appearance | Clear colorless to light yellow Liquid |
Infrared spectrum | Conforms |
Refractive index | 1.4810 to 1.4840 (20 °C, 589 nm) |
GC | ≥96.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...