4-(Diethylamino)salicylaldehyde 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 4-(Diethylamino)-2-hydroxybenzaldehyde

20948-0050 20948-0250
TS20948-0050EA 31 USD
TS20948-0050 TS20948-0250
4-(Diethylamino)salicylaldehyde 98%
4-(Diethylamino)salicylaldehyde
Formula: C₁₁H₁₅NO₂
MW: 193.25 g/mol
Melting Pt: 60…64 °C
Storage Temperature: Ambient
MDL Number: MFCD00003326
CAS Number: 17754-90-4
EINECS: 241-745-3

Order Now

Specification Test Results

Appearance Pink to light brown to purple Powder
Infrared spectrum Conforms
Melting point 60 to 64 °C
GC ≥97.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR