1,2-Diethoxyethane 99%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Diethyl glycol, Ethylene glycol diethyl ether

20824-0010 20824-0025 20824-1000
TS20824-0010EA 319 USD
TS20824-0010 TS20824-0025 TS20824-1000
1,2-Diethoxyethane 99%
1,2-Diethoxyethane
Formula: C₆H₁₄O₂
MW: 118.18 g/mol
Boiling Pt: 121.4 °C (1013 hPa)
Melting Pt: –74 °C
Density: 0.8417 g/cm³
Flash Pt: 19 °C
Storage Temperature: Ambient
MDL Number: MFCD00009253
CAS Number: 629-14-1
UN: 1153
ADR: 3,II

Order Now

Specification Test Results

Appearance Clear colorless Liquid
Infrared spectrum Conforms
Refractive index 1.3910 to 1.3930 (20 °C, 589 nm)
Water ≤0.1 %
GC ≥98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR