α-Terpinene 90% tech.

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: p-Mentha-1,3-diene, 1-Isopropyl-4-methylcyclohexa-1,3-diene

20749-1000 20749-5000
TS20749-1000EA 34 USD
TS20749-1000 TS20749-5000
α-Terpinene 90% tech.
α-Terpinene
Formula: C₁₀H₁₆
MW: 136.24 g/mol
Boiling Pt: 173 °C (1013 hPa)
Melting Pt: –48 °C
Density: 0.837 g/cm³
Flash Pt: 46 °C
MDL Number: MFCD00001534
CAS Number: 99-86-5
EINECS: 202-795-1
UN: 2319
ADR: 3,III

Order Now

Specification Test Results

Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.4770 to 1.4790 (20 °C, 589 nm)
Density (20°C) 0.832 to 0.842 g/ml
GC ≥88 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR