L-Leucic acid 98%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: (S)-2-Hydroxy-4-methylpentanoic acid, L-alpha-Hydroxyisocaproic acid, (S)-2-Hydroxy-4-methylvaleric Acid

20481-0010 20481-0050
TS20481-0010EA 52 USD
TS20481-0010 TS20481-0050
L-Leucic acid 98%
L-Leucic acid
Formula: C₆H₁₂O₃
MW: 132.16 g/mol
Melting Pt: 78…80 °C
Storage Temperature: Ambient
MDL Number: MFCD00064214
CAS Number: 13748-90-8

Order Now

Specification Test Results

Appearance White to off-white Crystalline powder
Infrared spectrum Conforms
Melting point 77 to 81 °C
NMR consistent with structure
Specific optical rotation -24° to -28° (20 °C, 589 nm) (C=1, 1N NaOH)
Titration with NaOH ≥97.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR