(±)-3-Octanol 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
3-Octanol, (±)-Ethyl pentyl carbinol, (±)-3-Hydroxy octane, (±)-Pentyl ethyl carbinol, DL-3-Octanol
20377-0500
20377-2500
TS20377-0500EA
49
USD
TS20377-0500
TS20377-2500
(±)-3-Octanol 97%
(±)-3-Octanol
Formula:
C₈H₁₈O MW: 130.23 g/mol Boiling Pt: 174…176 °C (760 mmHg) Melting Pt: –45 °C Density: 0.825 g/cm³ Flash Pt: 67 °C Storage Temperature: Ambient |
MDL Number:
MFCD00004590 CAS Number: 589-98-0 EINECS: 209-667-4 |
Specification Test Results
Appearance | Clear colorless Liquid |
Infrared spectrum | Conforms |
Refractive index | 1.4230 to 1.4320 (20 °C, 589 nm) |
Density | (20°C) 0.817 to 0.827 kg/l |
GC | ≥96.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...