(1R,4R)-Dihydrocarvone mixture of isomers 98%

Supplier: Thermo Scientific Chemicals

Synonyms: (2R,5R)-2-Methyl-5-(prop-1-en-2-yl)cyclohexanone

20365-0250 20365-1000
TS20365-0250EA 57 USD
TS20365-0250 TS20365-1000
(1R,4R)-Dihydrocarvone mixture of isomers 98%
(1R,4R)-Dihydrocarvone
Formula: C₁₀H₁₆O
Boiling Pt: 87…88 °C (6 mmHg)
Density: 0.9255 g/cm³
Flash Pt: 81 °C
Storage Temperature: Ambient
MDL Number: MFCD28125830
CAS Number: 5524-05-0

Order Now

Specification Test Results

Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.4700 to 1.4740 (20 °C, 589 nm)
GC ≥97.5 %
Specific gravity (25°C) 0.923 to 0.928
Specific optical rotation (20°C, 589 nm) (neat) > +14.0°


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR