(-)-Menthone 85%
Supplier: Thermo Scientific Chemicals
|
Warning
|
20361-0250
20361-1000
TS20361-0250EA
36
USD
TS20361-0250
TS20361-1000
(-)-Menthone 85%
(-)-Menthone
Formula:
C₁₀H₁₈O Flash Pt: 72 °C Storage Temperature: Ambient |
MDL Number:
MFCD00001634 CAS Number: 14073-97-3 EINECS: 237-926-1 |
Specification Test Results
Appearance | Clear colorless to light yellow Liquid |
Infrared spectrum | Conforms |
Refractive index | 1.4490 to 1.4530 (20 °C, 589 nm) |
GC | ≥80 % |
Specific optical rotation | -2° to -10° (20°C, 589 nm) (c=4, ethanol) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...