L(-)-Fenchone 98+%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: (1R)-(-)-Fenchone

18811-0500 18811-2500
TS18811-0500EA 45 USD
TS18811-0500 TS18811-2500
L(-)-Fenchone 98+%
L(-)-Fenchone
Formula: C₁₀H₁₆O
MW: 152.24 g/mol
Boiling Pt: 192 °C (1013 hPa)
Melting Pt: 5 °C
Density: 0.94 g/cm³ (0 °C)
Flash Pt: 52 °C
MDL Number: MFCD00151104
CAS Number: 7787-20-4
EINECS: 232-107-5
UN: 1224
ADR: 3,III

Order Now

Specification Test Results

Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.4600 to 1.4630 (20 °C, 589 nm)
GC ≥98.0 %
Specific optical rotation -47.5° to -53.5° (20°C, 589 nm) (neat)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR