3-Acetylthiophene 98%

Supplier: Thermo Scientific Chemicals
Warning

18802-0010 18802-0100
TS18802-0010EA 22 USD
TS18802-0010 TS18802-0100
3-Acetylthiophene 98%
3-Acetylthiophene
Formula: C₆H₆OS
Boiling Pt: 208…210 °C (748 mmHg)
Melting Pt: 57…61 °C
Storage Temperature: Ambient
MDL Number: MFCD00005468
CAS Number: 1468-83-3
EINECS: 215-996-4
UN: 2811
ADR: 6.1,III

Order Now

Specification Test Results

Appearance Beige to light brown Adhering crystals
Infrared spectrum Conforms
Melting point 57 to 61 °C
GC ≥97.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR