Thiophene-3-carbaldehyde 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Thiophene-3-carboxaldehyde, 3-Thiophenecarboxaldehyde

18798-0050 18798-0250 18798-1000
TS18798-0050EA 57 USD
TS18798-0050 TS18798-0250 TS18798-1000
Thiophene-3-carbaldehyde 98%
Thiophene-3-carbaldehyde
Formula: C₅H₄OS
MW: 112.15 g/mol
Boiling Pt: 194…196 °C (760 mmHg)
Melting Pt: –30 °C
Density: 1.28 g/cm³
Flash Pt: 64 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00005466
CAS Number: 498-62-4
EINECS: 207-865-5

Order Now

Specification Test Results

Appearance Clear yellow to light brown Liquid (May darken on storage)
Infrared spectrum Conforms
Refractive index 1.5820 to 1.5860 (20 °C, 589 nm)
GC ≥97.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR