(±)-6-Methyl-5-hepten-2-ol 99%

Supplier: Thermo Scientific Chemicals

18769-0250 18769-1000
TS18769-0250EA 79 USD
TS18769-0250 TS18769-1000
(±)-6-Methyl-5-hepten-2-ol 99%
(±)-6-Methyl-5-hepten-2-ol
Formula: C₈H₁₆O
Boiling Pt: 78 °C (14 mmHg)
Density: 0.84 g/cm³
Flash Pt: 67 °C
Storage Temperature: Ambient
MDL Number: MFCD00004561
CAS Number: 1569-60-4
EINECS: 216-377-1

Order Now

Specification Test Results

Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.4475 to 1.4495 (20 °C, 589 nm)
GC ≥98.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR