Cyanuric acid 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 1,3,5-Triazinetrion, 1,3,5-Triazine-2,4,6-triol, 1,3,5-Triazinane-2,4,6-trione, Isocyanuric acid

18102-0010 18102-0250 18102-2500
TS18102-0010EA 334 USD
TS18102-0010 TS18102-0250 TS18102-2500
Cyanuric acid 98%
Cyanuric acid
Formula: C₃H₃N₃O₃
Melting Pt: >360 °C
MDL Number: MFCD00082990
CAS Number: 108-80-5
EINECS: 203-618-0
Merck Index: 13,02728

Order Now

Specification Test Results

Appearance White to light beige Crystalline powder and/or lumps
Infrared spectrum Conforms
Titration with NaOH ≥97.5 %
GC ≥95.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR