1-Tetradecanol 99%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 1-Tetradecyl alcohol, 1-Hydroxy tetradecane, Myristyl alcohol

18073-0050 18073-0250 18073-1000 18073-5000
TS18073-0050EA 32 USD
TS18073-0050 TS18073-0250 TS18073-1000 TS18073-5000
1-Tetradecanol 99%
1-Tetradecanol
Formula: H₃C(CH₂)₁₂CH₂OH
MW: 214.39 g/mol
Boiling Pt: 289
Melting Pt: 37…40 °C
Flash Pt: 145 °C
Storage Temperature: Ambient
MDL Number: MFCD00004757
CAS Number: 112-72-1
EINECS: 204-000-3
UN: 3077
ADR: 9 III
Merck Index: 13,06361

Order Now

Specification Test Results

Appearance White Crystalline powder or crystals or flakes or pellets and/or chunks
Infrared spectrum Conforms
Melting point 37 to 41 °C
GC ≥98.5 %
Titration with HClO4 97.5 to 102.5 % (on dry substance)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR