4-Phenyl-1-butanol 97%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 4-Phenylbutan-1-ol

18037-0100 18037-0500
TS18037-0100EA 93 USD
TS18037-0100 TS18037-0500
4-Phenyl-1-butanol 97%
4-Phenyl-1-butanol
Formula: (C₆H₅)(CH₂)₃CH₂OH
MW: 150.22 g/mol
Boiling Pt: 140 °C (14 mmHg)
Density: 0.975 g/cm³
Storage Temperature: Ambient
MDL Number: MFCD00002971
CAS Number: 3360-41-6
EINECS: 222-128-8

Order Now

Specification Test Results

Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.5200 to 1.5220 (20 °C, 589 nm)
GC ≥96.0 %
Optical rotation +43° to +50° (1 % in water)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR