1-Adamantanemethanol 99%

Supplier: Thermo Scientific Chemicals

Synonyms: 1-(Hydroxymethyl)adamantane, 1-(Hydroxymethyl)tricyclo[3.3.1.1(3,7)]decane, Adamantan-1-ylmethanol, 1-Adamantylmethanol

18005-0025 18005-0100 18005-0500
TS18005-0025EA 38 USD
TS18005-0025 TS18005-0100 TS18005-0500
1-Adamantanemethanol 99%
1-Adamantanemethanol
Formula: C₁₁H₁₈O
MW: 166.26 g/mol
Melting Pt: 115…118 °C
Storage Temperature: Ambient
MDL Number: MFCD00074751
CAS Number: 770-71-8
EINECS: 212-225-3

Order Now

Specification Test Results

Appearance White Crystalline powder
Infrared spectrum Conforms
Melting point 115 to 118 °C
GC ≥98.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR