Methyl-2-chloroacetoacetate 95%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 2-Chloroacetoacetic acid methyll ester, Methyl 2-chloroacetoacetate

17990-0100 17990-0500
TS17990-0100EA 85 USD
TS17990-0100 TS17990-0500
Methyl-2-chloroacetoacetate 95%
Methyl-2-chloroacetoacetate
Formula: C₅H₇ClO₃
MW: 150.56 g/mol
Boiling Pt: 137 °C (1013 hPa)
Melting Pt: –33 °C
Density: 1.24 g/cm³
Flash Pt: 72 °C
Storage Temperature: Ambient
MDL Number: MFCD00008757
CAS Number: 4755-81-1
EINECS: 225-286-6
UN: 2810
ADR: 6.1,III

Order Now

Specification Test Results

Appearance Clear yellow Liquid
Infrared spectrum Conforms
Refractive index 1.4455 to 1.4475 (20 °C, 589 nm)
GC ≥94 %
HPLC ≥97.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR