2'-Chloroacetophenone 97%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 1-(2-Chlorophenyl)ethanone, o-Chloroacetophenone

17977-0250 17977-1000
TS17977-0250EA 46 USD
TS17977-0250 TS17977-1000
2'-Chloroacetophenone 97%
2'-Chloroacetophenone
Formula: C₈H₇ClO
MW: 154.6 g/mol
Boiling Pt: 227 °C (1013 hPa)
Density: 1.188 g/cm³
Flash Pt: 92 °C
Storage Temperature: Ambient
MDL Number: MFCD00000560
CAS Number: 2142-68-9
EINECS: 218-397-6

Order Now

Specification Test Results

Appearance Clear light yellow to amber Liquid (May darken on storage)
Infrared spectrum Conforms
Refractive index 1.5420 to 1.5450 (20 °C, 589 nm)
GC ≥96.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR