Methyl-6-quinolyl ether 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 6-Methoxyquinoline

17934-0250 17934-1000
TS17934-0250EA 81 USD
TS17934-0250 TS17934-1000
Methyl-6-quinolyl ether 98%
Methyl-6-quinolyl ether
Formula: C₁₀H₉NO
MW: 159.19 g/mol
Boiling Pt: 193 °C (50 mmHg)
Melting Pt: 18…20 °C
Density: 1.152 g/cm³
Flash Pt: >110 °C
Storage Temperature: Ambient
MDL Number: MFCD00006800
CAS Number: 5263-87-6
EINECS: 226-077-2

Order Now

Specification Test Results

Appearance Clear yellow to pink to purple to brown Liquid or low melting solid
Infrared spectrum Conforms
Refractive index 1.6220 to 1.6260 (20 °C, 589 nm)
GC ≥97.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR