Ethyl-4-chloroacetoacetate 98%
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
Ethyl 4-chloro-3-oxobutanoate
17764-0500
17764-2500
TS17764-0500EA
36
USD
TS17764-0500
TS17764-2500
Ethyl-4-chloroacetoacetate 98%
Ethyl-4-chloroacetoacetate
Formula:
C₆H₉ClO₃ MW: 164.59 g/mol Boiling Pt: 103 °C (16 mmHg) Melting Pt: –8 °C Density: 1.21 g/cm³ Flash Pt: 96 °C Storage Temperature: Ambient |
MDL Number:
MFCD00000939 CAS Number: 638-07-3 EINECS: 211-317-0 UN: 2927 ADR: 6.1,II |
Specification Test Results
Appearance | Clear colorless to yellow Liquid |
Infrared spectrum | Conforms |
Refractive index | 1.4515 to 1.4545 (20 °C, 589 nm) |
GC | ≥97.5 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...