trans,trans-2,4-Decadienal 95% tech.

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Decadienal, (2E,4E)-Deca-2,4-dienal, DDA

17751-0010 17751-0050 17751-0250
TS17751-0010EA 32 USD
TS17751-0010 TS17751-0050 TS17751-0250
trans,trans-2,4-Decadienal 95% tech.
trans,trans-2,4-Decadienal
Formula: C₁₀H₁₆O
MW: 152.24 g/mol
Boiling Pt: 114…116 °C (10 mmHg)
Density: 0.85 g/cm³
Flash Pt: 85 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00007007
CAS Number: 25152-84-5
EINECS: 246-668-9

Order Now

Specification Test Results

Appearance Clear yellow Liquid
Infrared spectrum Conforms
Refractive index 1.5140 to 1.5180 (20 °C, 589 nm)
GC ≥92 % (main isomer)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR