3-Quinolinecarbaldehyde 97+%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Quinoline-3-carboxaldehyde, Quinoline-3-carbaldehyde, 3-Quinolinecarboxaldehyde

17569-0010 17569-0050
TS17569-0010EA 116 USD
TS17569-0010 TS17569-0050
3-Quinolinecarbaldehyde 97+%
3-Quinolinecarbaldehyde
Formula: C₁₀H₇NO
MW: 157.17 g/mol
Melting Pt: 66…69 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00006768
CAS Number: 13669-42-6

Order Now

Specification Test Results

Appearance White to yellow Powder
Infrared spectrum Conforms
Melting point 66 to 69 °C
GC ≥97.0 %
HPLC ≥97.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR