1,1-Dimethoxyacetone 98%

Supplier: Thermo Scientific Chemicals
Warning

17380-1000 17380-5000
TS17380-1000EA 45 USD
TS17380-1000 TS17380-5000
1,1-Dimethoxyacetone 98%
1,1-Dimethoxyacetone
Formula: C₅H₁₀O₃
Boiling Pt: 143 °C (1013 hPa)
Melting Pt: –57 °C
Density: 1 g/cm³
Flash Pt: 37 °C
MDL Number: MFCD00008758
CAS Number: 6342-56-9
EINECS: 228-735-4
UN: 1224
ADR: 3,III

Order Now

Specification Test Results

Appearance Clear colorless to yellow Liquid
Infrared spectrum Conforms
Refractive index 1.3970 to 1.3990 (20 °C, 589 nm)
GC ≥97.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR