1-Docosanol 98%

Supplier: Thermo Scientific Chemicals

Synonyms: Behenyl alcohol

17372-0250 17372-1000
TS17372-0250EA 43 USD
TS17372-0250 TS17372-1000
1-Docosanol 98%
1-Docosanol
Formula: H₃C(CH₂)₂₀CH₂OH
MW: 326.61 g/mol
Boiling Pt: 376 °C (1013 hPa)
Melting Pt: 68…72 °C
Flash Pt: 227 °C
Storage Temperature: Ambient
MDL Number: MFCD00002939
CAS Number: 661-19-8
EINECS: 211-546-6
Merck Index: 13,03433

Order Now

Specification Test Results

Appearance White Pellets or tablets
Infrared spectrum Conforms
Melting point 68 to 72 °C
GC ≥97.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR