Ethyl-2-oxocyclopentanecarboxylate 95+%

Supplier: Thermo Scientific Chemicals

Synonyms: Ethylcyclopentanone-2-carboxylate, Ethyl 2-oxocyclopentanecarboxylate

17358-0250 17358-1000 17358-5000
TS17358-0250EA 46 USD
TS17358-0250 TS17358-1000 TS17358-5000
Ethyl-2-oxocyclopentanecarboxylate 95+%
Ethyl-2-oxocyclopentanecarboxylate
Formula: C₈H₁₂O₃
Boiling Pt: 224…228 °C (760 mmHg)
Density: 1.05 g/cm³
Flash Pt: 77 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00001412
CAS Number: 611-10-9
EINECS: 210-253-0

Order Now

Specification Test Results

Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.4500 to 1.4540 (20 °C, 589 nm)
GC ≥95.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR