3-Nitrobenzonitrile 98%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: m-Nitrobenzonitrile

17352-0050 17352-0250
TS17352-0050EA 24 USD
TS17352-0050 TS17352-0250
3-Nitrobenzonitrile 98%
3-Nitrobenzonitrile
Formula: C₇H₄N₂O₂
MW: 148.12 g/mol
Boiling Pt: 165 °C (21 mmHg)
Melting Pt: 115…119 °C
Storage Temperature: Ambient
MDL Number: MFCD00007194
CAS Number: 619-24-9
EINECS: 210-587-7
UN: 3439
ADR: 6.1,II

Order Now

Specification Test Results

Appearance Yellow Crystalline powder or needles
Infrared spectrum Conforms
Melting point 115 to 119 °C
GC ≥97.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR