Chloroacetone 96% stabilized

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Chloropropanone

17347-0010 17347-0050 17347-0250 17347-2500
TS17347-0010EA 115 USD
TS17347-0010 TS17347-0050 TS17347-0250 TS17347-2500
Chloroacetone 96% stabilized
Chloroacetone
Formula: C₃H₅ClO
MW: 92.52 g/mol
Boiling Pt: 120 °C (1013 hPa)
Melting Pt: –44.5 °C
Density: 1.16 g/cm³
Flash Pt: 32 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00000936
CAS Number: 78-95-5
EINECS: 201-161-1
UN: 1695
ADR: 6.1,I
Merck Index: 13,02131

Order Now

Specification Test Results

Appearance Clear yellow to yellow-brown Liquid
Infrared spectrum Conforms
Refractive index 1.4310 to 1.4330 (20 °C, 589 nm)
GC ≥95 %
Stabilizer 0.1 % epoxidized soybean oil

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR