2-Hydroxyisobutyric acid 98%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: 2-Hydroxy-2-methylpropionic acid, α-Hydroxyisobutyric acid, 2-Hydroxy-2-methylpropanoic acid

17310-0010 17310-0250 17310-1000
TS17310-0010EA 804 USD
TS17310-0010 TS17310-0250 TS17310-1000
2-Hydroxyisobutyric acid 98%
2-Hydroxyisobutyric acid
Formula: C₄H₈O₃
MW: 104.11 g/mol
Boiling Pt: 84 °C (1.5 mmHg)
Melting Pt: 77…81 °C
Storage Temperature: Ambient
MDL Number: MFCD00004459
CAS Number: 594-61-6
EINECS: 209-848-8

Order Now

Specification Test Results

Appearance White to cream Crystalline powder or crystals or needles
Infrared spectrum Conforms
Melting point 75 to 81 °C
Titration with NaOH ≥97.5 to 102.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR