2-(2-(2-Chloroethoxy)ethoxy)ethanol 95%
Supplier: Thermo Scientific Chemicals
|
Warning
|
17227-0100
17227-0500
TS17227-0100EA
50
USD
TS17227-0100
TS17227-0500
2-(2-(2-Chloroethoxy)ethoxy)ethanol 95%
2-(2-(2-Chloroethoxy)ethoxy)ethanol
Formula:
C₆H₁₃ClO₃ MW: 168.62 g/mol Boiling Pt: 256…258 °C (760 mmHg) Density: 1.163 g/cm³ Flash Pt: 107 °C Storage Temperature: Ambient |
MDL Number:
MFCD00002874 CAS Number: 5197-62-6 |
Specification Test Results
Appearance | Clear light yellow Liquid |
Infrared spectrum | Conforms |
GC | ≥94 % |
HPLC | ≥97.5 % |
Specific optical rotation | +158° to +168° (20°C, 589 nm) (c=1, water) On dry substance |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...