2-(2-Chloroethoxy)ethanol 99%

Supplier: Thermo Scientific Chemicals
Warning

17226-0500 17226-2500
TS17226-0500EA 60 USD
TS17226-0500 TS17226-2500
2-(2-Chloroethoxy)ethanol 99%
2-(2-Chloroethoxy)ethanol
Formula: C₄H₉ClO₂
MW: 124.57 g/mol
Boiling Pt: 79 °C (1013 hPa)
Density: 1.18 g/cm³
Flash Pt: 90 °C
MDL Number: MFCD00002870
CAS Number: 628-89-7
EINECS: 211-059-9
UN: 3334
ADR: 9,III

Order Now

Specification Test Results

Appearance Clear liquid
Infrared spectrum Conforms
Refractive index 1.4519 to 1.4539 (20 °C, 589 nm)
Water ≤1 % (K.F.)
GC ≥98.5 %
Color scale ≤20 APHA

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR