2-Amino-6-methoxybenzothiazole 98%

Supplier: Thermo Scientific Chemicals
Danger

17202-0050 17202-0500
TS17202-0050EA 22 USD
TS17202-0050 TS17202-0500
2-Amino-6-methoxybenzothiazole 98%
2-Amino-6-methoxybenzothiazole
Formula: C₈H₈N₂OS
Boiling Pt: 348 °C (1013 hPa)
Melting Pt: 162…169 °C
MDL Number: MFCD00005787
CAS Number: 1747-60-0
EINECS: 217-130-0
UN: 2811

Order Now

Specification Test Results

Appearance White to yellow to light beige Crystalline powder
Infrared spectrum Conforms
Melting point 165 to 169 °C
Titration with HClO4 ≥97.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR