3-Methyl-2-buten-1-ol 99%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: 2-Methyl-2-buten-4-ol

17192-0050 17192-1000 17192-5000
TS17192-0050EA 32 USD
TS17192-0050 TS17192-1000 TS17192-5000
3-Methyl-2-buten-1-ol 99%
3-Methyl-2-buten-1-ol
Formula: C₅H₁₀O
MW: 86.13 g/mol
Boiling Pt: 140 °C (1013 hPa)
Melting Pt: –86 °C
Density: 0.84 g/cm³
Flash Pt: 43 °C
Storage Temperature: Ambient
MDL Number: MFCD00002916
CAS Number: 556-82-1
EINECS: 209-141-4
UN: 1987
ADR: 3,III

Order Now

Specification Test Results

Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.4410 to 1.4450 (20 °C, 589 nm)
GC ≥98.5 %
Specific gravity (25°C) 0.857 to 0.867
Specific optical rotation -8.5° to -10.5° (20°C, 589 nm) (c=1, water)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR