3-Cyclohexylpropionic acid 99%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 3-Cyclohexylpropanoic acid, 3-Cyclohexylpropionic Acid

17145-0500 17145-2500
TS17145-0500EA 164 USD
TS17145-0500 TS17145-2500
3-Cyclohexylpropionic acid 99%
3-Cyclohexylpropionic acid
Formula: C₉H₁₆O₂
MW: 156.22 g/mol
Boiling Pt: 275.7 °C (1013 hPa)
Melting Pt: 14…17 °C
Density: 0.998 g/cm³
Flash Pt: 113 °C
Storage Temperature: Ambient
MDL Number: MFCD00001527
CAS Number: 701-97-3
EINECS: 211-861-9

Order Now

Specification Test Results

Appearance Clear colorless to light yellow Liquid (May darken on storage)
Infrared spectrum Conforms
Refractive index 1.4630 to 1.4650 (20 °C, 589 nm)
Titration with NaOH ≥98.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR